Carbon nanotube multi-walled core material

Supplier: STREM CHEMICALS INC MS
Warning

06-0505-1G 06-0505-5G
100207-326EA 247.69 USD
100207-326 100196-902
Carbon nanotube multi-walled core material
Carbon nanotube

Carbon Nanotubes, Nanomaterials

Formula: C
MDL Number: MFCD00133992
CAS Number: 308068-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR