3-Hydroxyphenylboronic acid pinacol ester ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

05-1043-5G 05-1043-1G
100201-316EA 610.03 USD
100201-316 100201-314
3-Hydroxyphenylboronic acid pinacol ester ≥97%
3-Hydroxyphenylboronic acid pinacol ester

Boronic Acid and Ester

Formula: C₁₂H₁₇BO₃
Melting Pt: 96…98 °C
MDL Number: MFCD02093755
CAS Number: 214360-76-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR