Medline® Electronic Blood Pressure Monitors
Supplier: MEDLINE INDUSTRIES, INC.
MDS9970 MDS9971 MDS9972 MDS9973 MDS3001 MDS3001PLUS
470303-262EA 24.35 USD
470303-262 470303-264 470303-266 470303-268 470303-272 470303-304 470303-306
Medline® Electronic Blood Pressure Monitors
Blood Pressure Monitors
Easy to use and adaptable.
  • Stores up to 90 measurements
  • One button testing
  • Talking version available
  • Interchangeable cuffs
These digital blood pressure monitors are an easy way to check a patients systolic pressure, diastolic pressure and pulse. Each monitor operates with a single button, automatically inflates and returns readings on a large easy to read digital face. Internal memory will retain the 90 most recent measurements as well as the date and time of each recording for easy tracking and rapid reference. Choose between the digital, with a larger base and talking option, or the more compact Elite digital. Each has the ability to change to larger or smaller cuffs (available separately) as desired.