Human Recombinant Neuritin-like protein
Supplier: Bon Opus Biosciences
CI43-10UG CI43-50UG CI43-500UG CI43-1MG
102943-834EA 107.69 USD
102943-834 102943-836 102943-838 102943-840
Human Recombinant Neuritin-like protein
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Neuritin-like protein is produced by our Mammalian expression system and the target gene encoding Ala36-Ala139 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.