Human Recombinant Tumor Necrosis Factor Receptor II
Supplier: Bon Opus Biosciences
CI39-10UG CI39-50UG CI39-500UG CI39-1MG
102943-802EA 118.46 USD
102943-802 102943-804 102943-806 102943-808
Human Recombinant Tumor Necrosis Factor Receptor II
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Tumor Necrosis Factor Receptor II is produced by our Mammalian expression system and the target gene encoding Leu23-Asp257 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.