Human Fam172A Ci26
Supplier: Bon Opus Biosciences
CI26-10UG CI26-1MG CI26-500UG CI26-50UG
102499-806EA 102.31 USD
102499-806 102499-808 102499-810 102499-812
Human Fam172A Ci26
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant Human Protein FAM172A is produced by our Mammalian expression system and the target gene encoding Gln19-Lys416 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx.
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.