Human Recombinant Sirpg
Supplier: Bon Opus Biosciences
CG10-10UG CG10-1MG CG10-500UG CG10-50UG
102865-166EA 107.69 USD
102865-166 102865-168 102865-170 102865-172
Human Recombinant Sirpg
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human signal-regulatory protein gamma is produced by our mammalian expression system and the target gene encoding Glu29-Pro360 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped via FedEx.
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.