Human Recombinant Sncg
Supplier: Bon Opus Biosciences
CF67-10UG CF67-1MG CF67-500UG CF67-50UG
102864-942EA 34.46 USD
102864-942 102864-944 102864-946 102864-948
Human Recombinant Sncg
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human gamma-synuclein is produced by our E.coli expression system and the target gene encoding Met1-Asp127 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped via FedEx.
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.