Human Recombinant Sult2B1
Supplier: Bon Opus Biosciences
CE93-10UG CE93-1MG CE93-500UG CE93-50UG
102864-438EA 107.69 USD
102864-438 102864-440 102864-442 102864-444
Human Recombinant Sult2B1
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human sulfotransferase 2B1 is produced by our E.coli expression system and the target gene encoding Met1-Glu311 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped via FedEx.
Packaging: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on dry ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.