Human Recombinant Znf100
Supplier: Bon Opus Biosciences
CE28-10UG CE28-1MG CE28-500UG CE28-50UG
102863-974EA 170.15 USD
102863-974 102863-976 102863-978 102863-980
Human Recombinant Znf100
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human zinc finger protein 100 is produced by our E.coli expression system and the target gene encoding Met1-Lys109 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped via FedEx.
Packaging: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on dry ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.