Human Recombinant Mp1
Supplier: Bon Opus Biosciences
CE21-10UG CE21-1MG CE21-500UG CE21-50UG
102863-926EA 170.15 USD
102863-926 102863-928 102863-930 102863-932
Human Recombinant Mp1
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human MAPKSP1 is produced by our E.coli expression system and the target gene encoding Met1-Val124 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped via FedEx.
Packaging: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on dry ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.