Human Recombinant Ilt1
Supplier: Bon Opus Biosciences
CA47-10UG CA47-1MG CA47-500UG CA47-50UG
102863-670EA 118.46 USD
102863-670 102863-672 102863-674 102863-676
Human Recombinant Ilt1
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human LILRA2 is produced by our mammalian expression system and the target gene encoding Gly24-Ser420 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx.
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.