Human Recombinant Carboxypeptidase B2
Supplier: Bon Opus Biosciences
CA30-10UG CA30-1MG CA30-500UG CA30-50UG
102863-622EA 170.15 USD
102863-622 102863-624 102863-626 102863-628
Human Recombinant Carboxypeptidase B2
Enzymes
It offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products.
  • Consistent sizing options
  • Datasheet included
  • High purity, low endotoxin levels
Recombinant human ribonuclease pancreatic is produced by our mammalian expression system and the target gene encoding Lys29-Thr156 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx.
Packaging: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on dry ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only. The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.