Human Recombinant Hr6A
Supplier: Bon Opus Biosciences
C183-10UG C183-1MG C183-500UG C183-50UG
102860-270EA 34.46 USD
102860-270 102860-272 102860-274 102860-276
Human Recombinant Hr6A
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Ubiquitin-Conjugating Enzyme E2 A is produced by our E.coli expression system and the target gene encoding Met1-Cys152 is expressed with a GST, 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.