Human Recombinant Tnf-Beta
Supplier: Bon Opus Biosciences
C181-10UG C181-1MG C181-500UG C181-50UG
102860-254EA 132.46 USD
102860-254 102860-256 102860-258 102860-260
Human Recombinant Tnf-Beta
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Tumor Necrosis Factor beta is produced by our E.coli expression system and the target gene encoding Leu35-Leu205 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.