Human Recombinant Dna Polymerase Beta
Supplier: Bon Opus Biosciences
C159-10UG C159-1MG C159-500UG C159-50UG
102860-078EA 118.46 USD
102860-078 102860-080 102860-082 102860-084
Human Recombinant Dna Polymerase Beta
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human DNA Polymerase beta is produced by our E.coli expression system and the target gene encoding Ser2-Glu335 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.