Human Recombinant G (from E. coli)
Supplier: Bon Opus Biosciences
C150-10UG C150-1MG C150-500UG C150-50UG
102860-014EA 118.46 USD
102860-014 102860-016 102860-018 102860-020
Human Recombinant G (from E. coli)
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Melanoma Inhibitory Activity Protein is produced by our E.coli expression system and the target gene encoding Gly25-Gln131 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.