Human Recombinant Carbonic Anhydrase 4
Supplier: Bon Opus Biosciences
C107-10UG C107-1MG C107-500UG C107-50UG
102859-678EA 67.85 USD
102859-678 102859-680 102859-682 102859-684
Human Recombinant Carbonic Anhydrase 4
Enzymes
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Carbonic Anhydrase 1 is produced by our E.coli expression system and the target gene encoding Ala2-Phe261 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9×6×7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.