Nickel 64% on silica stabilized, powder, reduced (Ni-5249P)

Supplier: STREM CHEMICALS INC MS
Danger

28-1900-250G 28-1900-50G
100207-646EA 1286.58 USD
100207-646 100209-888
Nickel 64% on silica stabilized, powder, reduced (Ni-5249P)
Nickel

Physical Characteristics: surface area 55m2/g

Formula: Ni
MDL Number: MFCD00011137
CAS Number: 7440-02-0
UN: 2881
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR