Anti-IgG F(ab)'2 Rabbit Polyclonal Antibody

Supplier: Immunoreagents
RB-005-C
10151-024EA 232.69 USD
10151-024
Anti-IgG F(ab)'2 Rabbit Polyclonal Antibody
Antibodies
  • Origin: US

Type:
Antigen: IgG F(ab)'2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR