Chloro{[(1S,2S)-(+)-2-amino-1,2-diphenylethyl](4-toluenesulfonyl)amido}(p-cymene)ruthenium(II) ≥90%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(S,S)-N-(p-Toluenesulfonyl)-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulphonyl)-1,2-diphenylethylenediamine]ruthenium(II), (S,S)-N-(p-Toluenesulphonyl)-1,2-diphenylethanediamine(chloro)(p-cymene)ruthenium(II), [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide]chloro(p-cymene)ruthenium(II), Chloro(p-cymene)[(S,S)-N-(p-toluenesulfonyl)-1,2-diphenylethylenediamine]ruthenium(II), [(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulfonamide]chloro(p-cymene)ruthenium(II), RuCl[(S,S)-Tsdpen](p-cymene)
Manufactured under license of Takasago patent US7129367B2. Takasago ATH Catalyst Kit component
Color and Form: yellow to dark brown solid
Stability: air sensitive
Formula:
C₃₁H₃₅ClN₂O₂RuS MW: 636.21 g/mol |
MDL Number:
MFCD09953484 CAS Number: 192139-90-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...