Withaferin A (from Withania somnifera), Calbiochem®

Supplier: MilliporeSigma

Synonyms: 5,6-Epoxy-4,27-dihydroxy-1-oxowitha-2,24-dienolide

681535-1MG 681535-5MG
EM681535-1MGEA 212.88 USD
EM681535-1MG EM681535-5MG
Withaferin A (from Withania somnifera), Calbiochem®
Withaferin A

Angiogenesis inhibitor.


  • White solid in form
  • Purity: ≥94% (by HPLC)


A cell-permeable steroidal lactone that displays antitumor, anti-inflammatory, radiosensitizing, and immunosuppressive properties. Potently inhibits angiogenesis both in vitro (IC50 = 12 nM against HUVEC proliferation) and in vivo (80% inhibition at 7 µg/kg/day in C57BL/6J mice, s.c.) and NF-κB activation (IC50 = 500 nM in TNF-α-induced endothelial cells) by targeting the ubiquitin-mediated proteasome pathway. At higher concentrations (~5 µM) also affects AP-1 transcription and induces cell death.


Isolated from Withania somnifera.


Packaging: Packaged under inert gas.

Formula: C₂₈H₃₈O₆
CAS Number: 5119-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR