Nutrient Agar Plates, Teknova
Supplier: Teknova
N0101 N0101 N0101
77443-106EA 130.66 USD
77443-106 CA100219-190 100219-190
Nutrient Agar Plates, Teknova
Media for Microbiology Dehydrated Media for Microbiology
This nutrient agar is used as a general purpose medium for the examination of water and dairy products. Also used for the cultivation of fastidious microorganisms.
It contains 0.3% Beef extract, 0.5% peptone, 0.8% NaCl and 1.5% agar.