Infusion Valves, Safsite® (B. Braun)
Supplier: B. Braun
413501
10821-480CS 353.84 USD
10821-480
Infusion Valves, Safsite® (B. Braun)
Infusions
Non-vented dispensing pin with two way Safsite® valve for injecting or withdrawing fluids from bags or semi-rigid containers.
  • Rapid fluid transfer with highest flow rate
  • Designed with larger lumen and high flow valves
  • Consist of self-sealing valve system
  • DEHP-free and latex-free