Anti-Angiotensin II Receptor 1 (AT1) Antibodies IgG (Rat) ELISA, Eagle Biosciences
Supplier: Eagle Biosciences
AT121-K01
102993-944EA 2818.08 USD
102993-944
Anti-Angiotensin II Receptor 1 (AT1) Antibodies IgG (Rat) ELISA, Eagle Biosciences
Assays ELISAs
The Rat Anti Angiotensin Receptor 1 (ATR1) IgG Antibody ELISA is an antibody screening test.
  • Sensitivity: Cut-off
  • Dynamic Range: 1.25 - 40 U/mL
  • Incubation Time: 3.5 hours
Angiotensin II is a peptide hormone which causes vasoconstriction, increased blood pressure, and release of aldosterone from the adrenal cortex. It occupies an important role in the ReninAngiotensin-System (RAS). Angiotensin II mediates the effects through G-Protein-coupled receptors, the Angiotensin receptors. The occurrence of autoantibodies against AT1 receptor is associated with an increased risk of an immunologic rejection after organ transplantation. The presence of AT1 autoantibodies correlate with the existence and course of Scleroderma. This Rat Anti Angiotensin II Receptor 1 (AT1R) IgG Antibody ELISA Assay Kit is designed for the determination of Rat antibodies (IgG) against the Angiotensin II receptor subtype I in serum and plasma.