Wangensteen Needle Holder, OR Grade, Sklar®
Supplier: Sklar
20-2780 20-2781
76192-528EA 423.75 USD
76192-528 76192-530
Wangensteen Needle Holder, OR Grade, Sklar®
Forceps
The Wangensteen needle nolder has pyramid-shaped teeth to reduce grip slippage when using fine needles.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
The straight jaws are to be used with 1-0, 2-0, and 3-0 sutures.