Glial-Mag, Transfection Reagent, OZ Biosciences
Supplier: OZ Biosciences
Glial-Mag
GL00250 KGL0250 GL00500
103537-342EA 889.28 USD
103537-342 103537-340 103537-338
Glial-Mag, Transfection Reagent, OZ Biosciences
Transfection Reagents
Glial-Mag is designed to reach optimal transfection efficiency with microglial cell lines and primary cells. Glial-Mag kit is the association of a specific magnetic nanoparticles formulation (Glial-Mag reagent), issued from our Magnetofection™ technology and a booster (Glial-Boost) designed to enhance transfection efficiency.
  • Highly efficient with microglial cell lines and primary cells
  • Low nucleic acid amount-minimized toxicity
  • High level of nucleic acid compaction
  • Easy and straightforward protocol
  • Compatible with any culture medium
Magnetofection™ is a highly efficient gene delivery method that exploits magnetic force exerted upon gene vectors associated with magnetic particles to drive the nucleic acids towards and into the target cells. The nucleic acid dose gets concentrated onto the cells within a few minutes and 100% of cells get in contact with a significant vector dose.
Delivery information: Shipped at room temperature.
Caution: Store at –20 °C.