Ammonium iron (II) sulfate hexahydrate ACS

Supplier: Pfaltz & Bauer
Warning

Synonyms: Ferrous ammonium sulfate hexahydrateAmmonium iron(Ⅱ) sulfate hexahydrateAmmonium ferrous sulphate hexahydratedi-Ammonium iron (II) sulphate hexahydrateAmmonium ferrous sulfate hexahydrateFerrous ammonium sulphate hexahydrateAmmonium iron(II) sulphate hydratedi-Ammonium iron (II) sulfate hexahydrateAmmonium iron(Ⅱ) sulphate hexahydrateAmmonium iron(II) sulfate hydrate

F01085-500G
101188-254EA 432.2 USD
101188-254
Ammonium iron (II) sulfate hexahydrate ACS
Ammonium iron (II) sulphate hexahydrate
Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Melting Pt: 100…110 °C
Density: 1.864 g/mL
MDL Number: MFCD00150530
CAS Number: 7783-85-9
EINECS: 233-382-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR