IncuCount Colony Counter

Supplier: REVOLUTIONARY SCIENCE SE
RS-IC-200
470235-876EA 7023 USD
470235-876
IncuCount Colony Counter
Colony Counters
IncuCount Automatic Colony Counter.

  • Counts up to 10 colony colors
  • Accommodates petri dishes up to 100 mm, including 3M® Petri film
  • Interactive software on 10" tablet (included)
  • USB flash drive data transfer port
  • Auto focus with override capabilities

The IncuCount Colony Counter has a high-quality HD digital camera featuring Carl Zeiss optics. The counter comes with a 10" tablet (android operating system) with a USB data transfer port for easy data storage. The unit has auto-focus with override capability, and performs colony counts and reads antibiotic reactions in a matter of seconds.

Delivery information: This item ships directly from the manufacturer.

Caution: For research or educational use only. Not for any human therapeutic or diagnostic use.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR