Dissolvable Paper Labels, Brady
Supplier: Brady
B30
THT-2250-403-WT THT-1125-403-WT B30C-1125-403-WT B30C-4000-403-WT THT-3000-403-WT THT-4000-403-WT B30C-2250-403-WT 149270
76193-284RL 140.02 USD
76193-284 76193-286 76193-290 76193-280 76193-282 76193-276 76193-288 76193-278
Dissolvable Paper Labels, Brady
Labels
This B-403 material completely dissolves in warm water within 30 seconds, leaving no adhesive residue behind.
  • Adhesive type: removable acrylic
  • Suggested ribbon series: R4300
  • Thickness: 0.0037"
Ideal for use in laboratory settings, asset tracking, or general-purpose use. Easy to remove, environmentally responsible, print-on-demand THT label solution. Easy removal saves time during re-labeling processes. Pressure-sensitive adhesive allows for quick and easy application.