Brady® i7100 Industrial Label Printers and Kit, Brady Worldwide
Supplier: Brady Worldwide
149050 149045 149056 149053 133251
76191-354EA 4941.88 USD
76191-354 76191-356 76191-358 76191-360 76250-754
Brady® i7100 Industrial Label Printers and Kit, Brady Worldwide
Printers Label Printers
Get the job done with the BradyPrinter i7100 label printer. No matter the demands, this reliable, heavy-duty and high-accuracy label printer gets it all done.
  • Single color printing capability
  • Display resolution: 480×272 pixels (H×W)
  • Display type: color touch screen
  • Power supply voltage: 100 to 40 V at 50 to 60 Hz
Print on the widest range of materials with confidence, including difficult to print materials. Prints ultra-sharp on labels as small as 0.125" with high accuracy to large labels with large text. Designed to print a maximum of 7000 labels perMore Product Information