Dehydrated Culture Media, YNB Media, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
114027522
114027532
114027512
114029622
76019-974EA
264.9
USD
76019-974
76020-282
IC114027512
76461-610
Dehydrated Culture Media, YNB Media, MP Biomedicals
Media for Microbiology
Dehydrated Media for Microbiology
YNB media, a component of synthetic defined yeast media is a well defined composition of salts, vitamins and nitrogen source for a vigorous growth of S. cerevisiae.