Riboprobe System-T7, 1 system, Promega
Supplier: Promega
P1440
PAP1440EA 248.64 USD
PAP1440
Riboprobe System-T7, 1 system, Promega
Nucleic Acid Reagents RNAi
The Riboprobe Systems are designed to prepare high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts.
  • Produce High-Specific-Activity RNA Probes or Defined RNA Transcripts
  • Systems available with SP6 RNA Polymerase, T7 RNA Polymerase or T3 RNA Polymerase
  • SP6, T7 and T3 RNA Polymerases are extremely promoter-specific, allowing production of virtually homogeneous RNA using plasmid DNA as a template
  • Allow synthesis of RNA corresponding to the coding or noncoding strand of cloned DNA from a single plasmid construct
The Riboprobe Systems are designed for in vitro preparation of high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts. These systems contain all components necessary for in vitro transcription from a DNA template (excluding the radioisotope) and also contain RQ1 RNase-Free DNase (Cat.# M6101) for template removal following transcription.
Caution: Store at –20°C