rGTP, 100 mM, 400 µl, Promega

Supplier: Promega Corporation

E6031 P1152
PAE6031EA 99.89 USD
PAE6031 PAP1152
rGTP, 100 mM, 400 µl, Promega
Nucleic Acid Reagents Nucleotides

High-quality rNTPs. Provided at concentrations of 10mM or 100mM in water at neutral pH.


  • High-Quality Ribonucleoside Triphosphates (rNTPs)
  • 400µl rNTP supplied at 10mM or 100mM in water at neutral pH
  • Qualified for use with the Riboprobe(R) and HeLaScribe(R) Systems
  • Purity verified by HPLC


Ribonucleotide Triphosphates (rNTPs) are provided in individual tubes and qualified for use with the Riboprobe and HeLaScribe Systems. The rNTPs are supplied in nuclease-free water. Purity is verified by HPLC analysis.

Order Now

SPECIFICATIONS

Environmentally Preferable
Application
Storage Conditions -30°C to -10°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR