PKA Kinase Enzyme System, 2,500 units, Promega
Supplier: Promega
V4246
PAV4246EA 624.55 USD
PAV4246
PKA Kinase Enzyme System, 2,500 units, Promega
Assays Enzyme Assays
Recombinant bovine cAMP-Dependent Protein Kinase (PKA), Catalytic Subunit, is expressed and purified from E. coli. PKA does not require cAMP for activity, and the gene sequence can be found at accession number NM_174584.
  • Easily Screen and Profile PKA Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Recombinant bovine cAMP-Dependent Protein Kinase (PKA), Catalytic Subunit, is expressed and purified from E. coli. Intracellular targets include ion channels (1), transcriptional activator proteins (2) and regulatory enzymes of glycogen metabolism (3). PKA does not require cAMP for activity, and the gene sequence can be found at accession number NM_174584. ADP-Glo Kinase Assay is a luminescent kinase assay that measures ADP formed from a kinase reaction; ADP is converted into ATP, which is a substrate in a reaction catalyzed by Ultra-Glo Luciferase that produces light. The luminescent signal positively correlates with ADP amount and kinase activity. The assay is well suited for measuring the effects chemical compounds have on the activity of a broad range of purified kinases, making it ideal for both primary screening as well as kinase selectivity profiling. The ADP-Glo Kinase Assay can be used to monitor the activity of virtually any ADP-generating enzyme (e.g., kinase or ATPase) using up to 1mM ATP. Kinase Enzyme System contains: Kinase: PKA, 2,500 units (Bovine, recombinant full-length). MW: ~40kDa. Substrate: Kemptide (LRRASLG). Other: Reaction Buffer, DTT. PKA NCBI Database Entry: www.ncbi.nlm.nih.gov/gene/282322/. Visit www.promega.com/kinase/ to see all Kinase Enzyme Systems.