YPG Agar, HiMedia
Supplier: HiMedia
M1368-500G M1368-2.5KG M1368-5KG
95021-028EA 211.85 USD
95021-028 89223-876 89223-878
YPG Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
YPG Agar is recommended for the growth of Saccharomyces cerevisiae for molecular biology purpose.
General methods in yeasts genetics specify using yeast extract, peptone medium for cultivating Saccharomyces cerevisiae
and other yeasts. The yeast extract allows faster growth so that during exponential or log-phase growth, the cells divide every
90 minutes. Peptic digest of animal tissue is as a source of carbon, nitrogen, vitamins and minerals.