Orthophosphoric acid 25% v/v
Supplier: AQUA SOLUTIONS MS
Synonyms:
Phosphoric acid
6867-500ML
6867-20L
6867-4L
6867-1L
101202-872EA
35.96
USD
101202-872
101203-612
101204-700
101203-490
Orthophosphoric acid 25% v/v
Orthophosphoric acid
Formula:
H₃PO₄ MW: 98 g/mol Boiling Pt: 158 Melting Pt: 21 |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 UN: 1805 ADR: 8 III Merck Index: 14,07344 |
Specification Test Results
Purity (v/v) | 25% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...