Potassium hydrogen diiodate 0.025 N
Supplier: AQUA SOLUTIONS MS
Synonyms:
Potassium bi-iodate
6895-4L
6895-1L
101204-118EA
160.08
USD
101204-118
101204-898
Potassium hydrogen diiodate 0.025 N
Potassium hydrogen diiodate
Formula:
HI₂KO₆ MW: 389.91 g/mol |
MDL Number:
MFCD00011400 CAS Number: 13455-24-8 |
Specification Test Results
Normality | 0.025 N |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...