2,3-Dichloro-5,6-dicyano-p-benzoquinone ≥98.5%, yellow crystalline powder

Supplier: CHEM IMPEX INTL INC MS
Danger

Synonyms: DDQ2,3-Dichloro-5,6-dicyanoquinone2,3-Dichloro-5,6-dicyano-1,4-benzoquinone4,5-Dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile

21905-5KG 21905-100G 21905-25G 21905-250G 21905-1KG
102623-904EA 69.86 USD
102623-904 102553-416 102561-810 102568-610 102598-412
2,3-Dichloro-5,6-dicyano-p-benzoquinone ≥98.5%, yellow crystalline powder
2,3-Dichloro-5,6-dicyano-p-benzoquinone

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone is used as an oxidant for the dehydrogenation of alcohol, phenol, steroid ketones and in the synthesis of quinolines from imines and alkynes or alkenes. It is a deprotection reagent for thioacetals, acetals and ketals.

Formula: C₈Cl₂N₂O₂
MW: 227.01 g/mol
Melting Pt: 214.0…218.0 °C
Storage Temperature: Freezer
MDL Number: MFCD00001593
CAS Number: 84-58-2
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Infrared Spectrum Conforms to Structure
Loss on Drying ≤0.5%
Residue On Ignition ≤0.1%
Assay (Titration) ≥98.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR