Magnesium chloride hexahydrate 99.0-102.0% ACS, Fluka™
Supplier: Honeywell Research Chemicals
M9272-100G
M9272-1KG
M9272-2.5KG
M9272-4X2.5KG
M9272-500G
M9272-6X500G
BJM9272-100GEA
65
USD
BJM9272-100G
BJM9272-1KG
BJM9272-2.5KG
BJM9272-4X2.5
BJM9272-500G
BJM9272-6X500G
Magnesium chloride hexahydrate 99.0-102.0% ACS, Fluka™
Magnesium chloride hexahydrate
Formula:
MgCl₂·6H₂O Melting Pt: 117 °C (–H₂O of crystall.) Density: 1.57 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149781 CAS Number: 7791-18-6 EINECS: 232-094-6 |
Specification Test Results
Assay | 99.0 - 102.0 % |
Water insoluble matter | Max. 0.005 % |
Barium (Ba) | Max. 0.005 % |
Calcium (Ca) | Max. 0.01 % |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 0.005 % |
Manganese (Mn) | Max. 5 ppm |
Sodium (Na) | Max. 0.005 % |
Strontium (Sr) | Max. 0.005 % |
Heavy metals (as Pb) | Max. 5 ppm |
Nitrate (NO₃) | Max. 0.001 % |
Phosphate (PO₄) | Max. 5 ppm |
Sulfate (SO₄) | Max. 0.002 % |
Ammonium (NH₄) | Max. 0.002 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...