L-Ornithine methyl ester dihydrochloride 99%, white crystalline powder

Supplier: CHEM IMPEX INTL INC MS

Synonyms: H-Orn-OMe.2HCl, H-Orn-OMe·2 HCl, (S)-methyl 2,5-Diaminopentanoate dihydrochloride, H-Orn-OMe·2HCl

06154-5G 06154-100G 06154-250G 06154-25G
102592-194EA 27.08 USD
102592-194 102592-196 102592-198 102632-370
L-Ornithine methyl ester dihydrochloride 99%, white crystalline powder
L-Ornithine methyl ester dihydrochloride
Formula: C₆H₁₆Cl₂N₂O₂
MW: 219.106 g/mol
Melting Pt: 180…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190977
CAS Number: 40216-82-8

Order Now

Specification Test Results

Optical Rotation, [a]D30 +11 ±2° (C=1 in MeOH)
Purity ≥98%
Melting Point 180 - 188 ºC


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR