DuPont™ Tychem® 5000 Sleeved Aprons
Supplier: DuPont
Tychem®
C3275TTNLG000600 C3275TTNXL000600 C3275TTNMD000600 C3275TTN3X000600 C3275TTN2X000600 C3275TTN4X000600 C3275TTNSM000600
76187-408CS 225.35 USD
76187-408 76187-398 76187-410 76187-400 76187-402 76187-404 76187-406
DuPont™ Tychem® 5000 Sleeved Aprons
Disposable Garments
Tychem® 5000 fabric is composed of a multi-layer film barrier laminated to a durable 2.0 oz/yd2 polypropylene substrate.
  • Sleeved
  • Elastic wrists
  • Neck loop with snaps and waist ties
  • Taped seams
Tychem® 5000 fabric is strong and durable for rigorous activities and rugged situations involving liquid splash and provides barrier to a broad range of chemicals. Typical Applications: chemical handling, petro-chemical, hazardous materials/waste clean-up, fire departments, industrial hazmat teams, utilities, and domestic preparedness. Commonly used in domestic preparedness for situations where the potential to exposure to chemicals exist. These aprons are tan.
These aprons are knee-length gowns and feature attached sleeves, which provide enhanced frontal protection. Open back allows for wearer comfort. Bound seams around neck opening feature a snap closure. Aprons also feature ties at the waist and elastic openings at wrists for a tighter fit.
Certifications: Thickness meets ASTM D1777.