Anti-SILV Mouse Monoclonal Antibody (CF568) [clone: NKI-beteb]

Supplier: Biotium

BNC680226-500 BNC680226-100
75970-752EA 197.58 USD
75970-752 75970-946
Anti-SILV Mouse Monoclonal Antibody (CF568) [clone: NKI-beteb]
Antibodies
By immunohistochemistry this antibody specifically recognizes a in melanocytes and melanomas. This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin.This Mab labels formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation.

CF® dyes are Biotium's next-generation fluorescent dyes. CF®568 is a red fluorescent dye (Ex/Em 562/583 nm) with superior brightness and photostability. It also is compatible with super-resolution imaging by STORM and TIRF.

Type: Primary
Antigen: SILV
Clonality: Monoclonal
Clone: Clone NKI-beteb
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Horse, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR