Anti-TAG-72 Mouse Monoclonal Antibody [clone: CA72/145]

Supplier: Biotium

BNUB0145-100 BNUB0145-500
75974-746EA 240.92 USD
75974-746 75974-748
Anti-TAG-72 Mouse Monoclonal Antibody [clone: CA72/145]
Antibodies
This antibody recognizes an oncofetal antigen of 220 kDa, identified as a tumor-associated glycoprotein (TAG-72) with properties of a mucin. The majority of human adenocarcinomas including colorectal, pancreatic, gastric, ovarian, endometrial, mammary, and non-small cell lung cancer display some cell populations that are positive for TAG72. About 60% of carcinoma patients express TAG72 in their sera. TAG72 is reportedly useful in distinguishing pulmonary adenocarcinomas from pleural mesotheliomas.

Type: Primary
Antigen: TAG-72
Clonality: Monoclonal
Clone: Clone CA72/145
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR