Ammonium iron (III) sulfate 0.1 M (NH4)Fe(SO4)2 volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Ammonium ferric sulphateFerric ammonium sulfateFerric ammonium sulphateAmmonium ferric sulfate

34249-1L
BJ34249-1LEA 169.62 USD
BJ34249-1L
Ammonium iron (III) sulfate 0.1 M (NH4)Fe(SO4)2 volumetric solution, Fluka™
Ammonium iron (III) sulphate
Formula: H₄FeNO₈S₂
MW: 266.01 g/mol
Density: 1.05 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00012019
CAS Number: 10138-04-2
UN: 3264
ADR: 8,III

Order Now

Specification Test Results

Factor 0.998 - 1.002

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR