Chloramphenicol 34 mg/mL in ethanol 100%

Supplier: TEKNOVA INC

Synonyms: Chloromycetin®(R,R)-ChloramphenicolTega-Cetin

C0322
101319-572EA 53.88 USD
101319-572
Chloramphenicol 34 mg/mL in ethanol 100%
Chloramphenicol

Chloramphenicol is a broad-spectrum antibiotic effective against most strains of Gram-positive and Gram-negative bacteria. It is bacteriostatic but may be bactericidal at high concentrations and is lipid-soluble, allowing it to diffuse through the bacterial cell membrane.

Formula: Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
MW: 323.13 g/mol
MDL Number: MFCD00078159
CAS Number: 56-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR