x-tracta™ Gel Extractor, Promega
Supplier: Promega Corporation
A2121 A2122
PAA2121EA 32.45 USD
PAA2121 PAA2122
x-tracta™ Gel Extractor, Promega
Cutters
The x-tracta Gel Extractor tool provides a convenient, safe method for removal of agarose gel fragments for further processing. The device removes a 0.13 x 0.33 inch gel piece from agarose gels for easy transfer into a microcentrifuge tube for processing.
  • Safe, Convenient Extraction of Gel Slices
  • Removes a 0.13 x 0.33 inch gel piece
  • Disposable device reduces cross-contamination
  • No need to use razor blades or scalpels
The x-tracta Gel Extractor tool provides a convenient, safe method for removal of agarose gel fragments for further processing. The device removes a 0.13 x 0.33 inch gel piece from agarose gels for easy transfer into a microcentrifuge tube for processing. The x-tracta tool eliminates the need for razor blades or scalpels, and its single-use design eliminates the possibility for sample-to-sample cross-contamination. Note: The x-tracta Gel Extractor works best on 0.6-2% analytical grade agarose gels. Please exercise caution if using the x-tracta Gel Extractor on Low Melting Point (LMP) agarose gels because the extractor does not work effectively due to the gel consistency.