COD Calibration Standard Standards
Supplier: CHEMetrics
A-7301 A-7310
CXA-7301EA 38.11 USD
CXA-7301 CXA-7310
COD Calibration Standard Standards
Standards Environmental Analysis Standards
COD calibration standards are designed to be run as a known standard alongside test vials to verify that your test procedure was performed correctly.
  • Reliable COD standard
  • Convenient 200 ml volume
  • Packaged in brown glass bottle
  • Chemical method: Dichromate reactor digestion
COD calibration standards are designed to be run as a known standard alongside test vials to verify that your test procedure was performed correctly. Prevent mistakes or inaccurate results by conducting a known standard alongside your tests.
Caution: This standard must be refrigerated.