Protecta Compliance in a Can™ Light Roofer's Fall Protection Kit, ORS Nasco
Supplier: ORS Nasco
098-2199802
76111-206EA 122.64 USD
76111-206
Protecta Compliance in a Can™ Light Roofer's Fall Protection Kit, ORS Nasco
Fall Protection Fall Protection Kits
  • Complete all-purpose fall protection system in one handy container
  • Durable bucket with handle
Includes lightweight FIRST™ harness with pass-thru buckle legs and 5-point adjustment (model 1191995) for easy wear. Sturdy PRO™ 6' (1.8m) single-leg shock absorbing lanyard (model 1341001) ensures security.