DBI-SALA® ExoFit NEX™ Black-Out RAR Harnesses, ORS Nasco
Supplier: Safety & Industrial Supplies
098-1113370 098-1113371 098-1113372
76111-306EA 653.63 USD
76111-306 76111-308 76111-310
DBI-SALA® ExoFit NEX™ Black-Out RAR Harnesses, ORS Nasco
Fall Protection Fall Harnesses
Repel™ Web and Tech-Lite™ resist water, reducing the attraction of mold and dirt. D-rings are aluminum on the front, back, and sides.
  • Hybrid comfort padding provides a lightweight, moisture wicking, and breathable padding that will not move or slip, always keeping the worker comfortable
  • Duo-Lok™ quick-connect buckles
  • Easy holstered snap hooks will break away to avoid trip hazard
  • Superior X-Design has a unique X construction that wraps around the user for the ultimate in comfort, security, and no-tangle donning
  • Ergonomically lightweight leg padding provides greater comfort for longer use
For prolonged hours of usage, an ergonomic body belt and pad offer increased comfort and choices for work positions. Three equipment loops with protective sheathing make it simple to access additional tools and gear.
Certifications: Certified to NFPA 1983 Class III, Meets OSHA, ANSI Z359.1.